Learn More
Biomatik Corporation Recombinant Matrix Metalloproteinase 3 (MMP3), 100UG
Supplier:  Biomatik Corporation RPU42713
Encompass_Preferred
Recombinant Rat Matrix Metalloproteinase 3 (MMP3) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Matrix Metalloproteinase 3 (MMP3). Target Synonyms: SL1; STMY; STMY1; STR1; Progelatinase; Stromelysin 1; Transin-1. Accession Number: P03957. Expression Region: Asn278~Gly450. Tag Info: N-terminal His Tag. Theoretical MW: 20kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5657
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.