Learn More
Biomatik Corporation Recombinant Parathyroid Hormone (PTH), 100UG
Supplier:  Biomatik Corporation RPU43180
Encompass_Preferred
Recombinant Human Parathyroid Hormone (PTH) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Parathyroid Hormone (PTH). Target Synonyms: iPTH; Intact Parathyroid Hormone; Parathormone; Parathyrin. Accession Number: P01270. Gene ID: 5741. Expression Region: Arg31~Gln115. Tag Info: N-terminal His Tag. Theoretical MW: 13kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5691
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.