Learn More
Biomatik Corporation Recombinant Perforin 1 (PRF1), 100UG
Supplier:  Biomatik Corporation RPU43217
Encompass_Preferred
Recombinant Mouse Perforin 1 (PRF1) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Perforin 1 (PRF1). Target Synonyms: FLH2; HPLH2; P1; PFP; Pore Forming Protein; Cytolysin; Lymphocyte pore-forming protein. Accession Number: P10820. Gene ID: 18646. Expression Region: Val40~Lys355. Tag Info: N-terminal His and GST Tag. Theoretical MW: 65kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5696
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.