Learn More
Biomatik Corporation Recombinant Tumor Necrosis Factor Related Apoptosis Inducing Ligand (TRAIL), 100UG
Supplier:  Biomatik Corporation RPU44262
Encompass_Preferred
Recombinant Rat Tumor Necrosis Factor Related Apoptosis Inducing Ligand (TRAIL) is a purified Prokaryotic Protein. Purity: >80% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Tumor Necrosis Factor Related Apoptosis Inducing Ligand (TRAIL). Target Synonyms: CD253; TNFSF10; APO2L; Apo2-L; TL2; TRAIL; Tumor Necrosis Factor Ligand Superfamily Member 10; Apo-2 Ligand. Accession Number: E9PU84. Expression Region: Pro115~Ala287. Tag Info: N-terminal His and GST Tag. Theoretical MW: 50kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5771
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.