Learn More
Biomatik Corporation Native Advanced Glycation End Product (AGE), 100UG
Supplier:  Biomatik Corporation RPU50405
Encompass_Preferred
Native Advanced Glycation End Product (AGE) is a purified Natural Protein. Purity: >90% by SDS-PAGE. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Universal. Target Name: Advanced Glycation End Product (AGE). Target Synonyms: AGEs; Advanced Glycation End Products. Buffer: PBS, pH7.4, containing 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5795
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.