Learn More
Biomatik Corporation BSA Conjugated Estradiol (E2), 100UG
Supplier:  Biomatik Corporation RPU50787
Encompass_Preferred
BSA Conjugated Estradiol (E2) is a purified Conjugated Small Molecules. Purity: >90% by SDS-PAGE. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Universal. Target Name: Estradiol (E2). Target Synonyms: 17B-Estradiol; Oestradiol; Beta-Estradiol. Tag Info: N-terminal His Tag. Theoretical MW: 272.4Da. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5804
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.