Learn More
Biomatik Corporation Recombinant Procollagen III N-Terminal Propeptide (PIIINP), 100UG
Supplier:  Biomatik Corporation RPU51440
Encompass_Preferred
Recombinant Human Procollagen III N-Terminal Propeptide (PIIINP) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Procollagen III N-Terminal Propeptide (PIIINP). Target Synonyms: P3NP; N-Propeptide Of Type III Procollagen; Procollagen III Amino Terminal Propeptide. Accession Number: P02461. Gene ID: 1281. Expression Region: Gln24~Pro153. Tag Info: Two N-terminal Tags, His-tag and SUMO-tag. Theoretical MW: 38kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5819
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.