Learn More
Biomatik Corporation Recombinant High Mobility Group Protein 17 (HMG17), 100UG
Supplier:  Biomatik Corporation RPU54468
Encompass_Preferred
Recombinant Human High Mobility Group Nucleosome Binding Domain Containing Protein 2 (HMGN2) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: High Mobility Group Nucleosome Binding Domain Containing Protein 2 (HMGN2). Target Synonyms: HMG17; High Mobility Group Protein 17; Non-histone chromosomal protein HMG-17. Accession Number: P05204. Gene ID: 3151. Expression Region: Pro2~Gln81. Tag Info: N-terminal His Tag. Theoretical MW: 17kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5905
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.