Learn More
Biomatik Corporation Active Mannose Associated Serine Protease 2 (MASP2), 100UG
Supplier:  Biomatik Corporation RPU54833
Encompass_Preferred
Human Mannose Associated Serine Protease 2 (MASP2), Active Protein is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Mannose Associated Serine Protease 2 (MASP2). Target Synonyms: Mannose-binding protein-associated serine protease 2; mannan-Binding Lectin Serine Peptidase 2; MBL-associated serine protease 2. Accession Number: O00187. Gene ID: 10747. Expression Region: Ile445~Ile683. Tag Info: N-terminal His Tag. Theoretical MW: 27kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5925
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.