Learn More
Biomatik Corporation Recombinant Escherichia coli O6:H1 Cell division protein FtsZ (ftsZ), 100UG
Supplier:  Biomatik Corporation RPC22729
Encompass_Preferred
Recombinant Escherichia coli O6:H1 Cell division protein FtsZ (ftsZ) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Escherichia coli O6:H1 (strain CFT073 / ATCC 700928 / UPEC). Target Name: ftsZ. Target Synonyms: ftsZ; c0113Cell division protein FtsZ. Accession Number: P0A9A7. Expression Region: 1~383aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 56.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5176
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.