Learn More
Biomatik Corporation Recombinant Bone Morphogenetic Protein 2 (BMP2), 100UG
Supplier:  Biomatik Corporation RPU40486
Encompass_Preferred
Recombinant Human Bone Morphogenetic Protein 2 (BMP2) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Bone Morphogenetic Protein 2 (BMP2). Target Synonyms: BMP2A; BMP-2A; Hemochromatosis Modifier. Accession Number: P12643. Gene ID: 650. Expression Region: Gln283~Arg396. Tag Info: N-terminal His and GST Tag. Theoretical MW: 43kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5484