Learn More
Biomatik Corporation Recombinant Human Cancer/testis antigen 1 (CTAG1A)
Supplier:  Biomatik Corporation RPC21152
Encompass_Preferred
Recombinant Human Cancer/testis antigen 1 (CTAG1A) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: CTAG1A. Target Synonyms: Autoimmunogenic cancer/testis antigen NY-ESO-1Cancer/testis antigen 6.1. Accession Number: P78358. Expression Region: 1~180aa. Tag Info: N-Terminal 10Xhis-Tagged. Theoretical MW: 21.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0460
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.