Learn More
Biomatik Corporation Recombinant Human Proteoglycan 4 (PRG4)
Supplier:  Biomatik Corporation RPU43521
Encompass_Preferred
Recombinant Human Proteoglycan 4 (PRG4) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Proteoglycan 4 (PRG4). Target Synonyms: MSF; CACP; HAPO; JCAP; SZP; Megakaryocyte Stimulating factor; Articular Superficial Zone Protein; Camptodactyly; Arthropathy,Coxa Vara,Pericarditis Syndrome. Accession Number: Q92954. Gene ID: 10216. Expression Region: Val1151~Gly1241. Tag Info: N-terminal His Tag. Theoretical MW: 12kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0050