Learn More
Biomatik Corporation Recombinant Mouse Cyclin Dependent Kinase Inhibitor 2A (CDKN2A)
Supplier:  Biomatik Corporation RPU41087
Encompass_Preferred
Recombinant Mouse Cyclin Dependent Kinase Inhibitor 2A (CDKN2A) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Cyclin Dependent Kinase Inhibitor 2A (CDKN2A). Target Synonyms: P16; ARF; CDK4I; CMM2; INK4; INK4a; MLM; MTS1; TP16; P14; P14ARF; P16INK4a; P19; Melanoma P16; Cyclin-dependent kinase 4 inhibitor A; Multiple tumor suppressor 1. Accession Number: P51480. Gene ID: 12578. Expression Region: Ser3~Thr154. Tag Info: N-terminal His Tag. Theoretical MW: 20kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9918
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.