Learn More
Biomatik Corporation Recombinant Rat Macrophage Migration Inhibitory Factor (MIF)
Supplier:  Biomatik Corporation RPU42656
Encompass_Preferred
Recombinant Rat Macrophage Migration Inhibitory Factor (MIF) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Macrophage Migration Inhibitory Factor (MIF). Target Synonyms: GIF; GLIF; MMIF; Glycosylation-Inhibiting Factor; L-dopachrome isomerase; L-dopachrome tautomerase; Phenylpyruvate tautomerase. Accession Number: P30904. Gene ID: 81683. Expression Region: Phe4~Ser112. Tag Info: N-terminal His Tag. Theoretical MW: 14kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0013
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.