Learn More
Biomatik Corporation Recombinant Human Procollagen C-Endopeptidase Enhancer (PCOLCE)
Supplier:  Biomatik Corporation RPU43418
Encompass_Preferred
Recombinant Human Procollagen C-Endopeptidase Enhancer (PCOLCE) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Procollagen C-Endopeptidase Enhancer (PCOLCE). Target Synonyms: PCPE1; PCPE; Procollagen Type 1 COOH-Terminal Proteinase Enhancer; Procollagen C Endopeptidase Enhancer; Type I procollagen COOH-terminal proteinase enhancer; Procollagen C Proteinase Enhancer 1. Accession Number: Q15113. Gene ID: 5118. Expression Region: Gly38~Asp449. Tag Info: N-terminal His Tag. Theoretical MW: 48kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0045
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.