Learn More
Biomatik Corporation Recombinant Human Cyclin Dependent Kinase 9 (CDK9)
Supplier: Biomatik Corporation RPU52524
Recombinant Human Cyclin Dependent Kinase 9 (CDK9) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Cyclin Dependent Kinase 9 (CDK9). Target Synonyms: C-2k; CDC2L4; PITALRE; TAK; CDC2-Related Kinase; Cell division cycle 2-like protein kinase 4; Cell division protein kinase 9; Tat-associated kinase complex catalytic subunit. Accession Number: P50750. Gene ID: 1025. Expression Region: Asp104~Phe315. Tag Info: N-terminal His Tag. Theoretical MW: 28kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.