Learn More
Biomatik Corporation Recombinant Human Colony Stimulating Factor 2, Granulocyte Macrophage (GM-CSF)
Supplier:  Biomatik Corporation RPU40924
Encompass_Preferred
Recombinant Human Colony Stimulating Factor 2, Granulocyte Macrophage (GM-CSF) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Colony Stimulating Factor 2, Granulocyte Macrophage (GM-CSF). Target Synonyms: CSF2; GMCSF; Sargramostim; Molgramostin; Granulocyte-Macrophage Colony Stimulating Factor. Accession Number: P04141. Gene ID: 1437. Expression Region: Ala18~Glu144. Tag Info: N-terminal His and GST Tag. Theoretical MW: 45kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9905
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.