Learn More
Biomatik Corporation Synthetic Human Oxidized Low Density Lipoprotein (OxLDL)
Supplier:  Biomatik Corporation RPU51037
Encompass_Preferred
Synthetic Human Oxidized Low Density Lipoprotein (OxLDL) is a purified Synthetic Peptides. Purity: >95% by SDS-PAGE. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Oxidized Low Density Lipoprotein (OxLDL). Target Synonyms: OL. Accession Number: P04114. Gene ID: 338. Tag Info: N-terminal His Tag. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0120
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.