Learn More
Biomatik Corporation Recombinant Human Surfactant Associated Protein A2 (SPA2)
Supplier:  Biomatik Corporation RPU54440
Encompass_Preferred
Recombinant Human Surfactant Associated Protein A2 (SPA2) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Surfactant Associated Protein A2 (SPA2). Target Synonyms: SFTPA2B; COLEC5; SP2A; SPA1 beta; SP-A2; SPAII; Collectin-5; Pulmonary Surfactant Protein A2; 35 kDa pulmonary surfactant-associated protein; Alveolar proteinosis protein. Accession Number: Q8IWL1. Gene ID: 729238. Expression Region: Glu21~Phe114. Tag Info: N-terminal His and GST Tag. Theoretical MW: 41kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0244
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.