Learn More
Biomatik Corporation Recombinant Human Collagen alpha-1 (IV) chain (COL4A1), partial
Supplier: Biomatik Corporation RPC21120
Recombinant Human Collagen alpha-1 (IV) chain (COL4A1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: COL4A1. Target Synonyms: Arresten; BSVD; CO4A1_HUMAN; COL4A1; COL4A1 NC1 domain; COL4A2; COL4A3; COL4A4; COL4A5; collagen alpha-1(IV) chain; Collagen IV Alpha 1 Polypeptide; Collagen IV Alpha 2 Polypeptide. Accession Number: P02462. Expression Region: 1445~1669aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 40.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.