Learn More
Bachem Cyclo(-His-Phe)
SDP
Supplier:  Bachem 4006752.1000
Encompass_Preferred
Cyclo(-His-Phe), 56586-95-9, C15H16N4O2, Mr 284.32, 1g. Catalyst for asymmetric cyanohydrin synthesis. The addition of hydrogen cyanide to benzaldehyde gives the highest optical yield ever obtained for the corresponding cyanohydrin. In a study of McCleland et al., Cyclo(His-Phe) showed significant anti-tumour activity, causing reduction of cell viability in cervical cancer cells. In isolated rat hearts, the diketopiperazine caused a gradual reduction of heart rate and a decrease in coronary flow rate.
Catalog No. 50-194-6705
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.