Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Catch Tubes

Catalog No. MP116560211 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP116560211 50 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116560211 Supplier MP Biomedicals™ Supplier No. 116560211
Add to Cart
Add to Cart

Catch Tubes are used during the silica-based purification procedure to eliminate accidental transfer of silica particles.

  • Included in FastDNA SPIN Kits and in the GENECLEAN SPIN Kit
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Quantity 50 Pk.
Product Type Catch Tubes
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.