Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Catechin 5-O-gallate, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012908 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2908 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2908 Supplier MilliporeSigma Supelco Supplier No. 8910610MG
Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 128232-62-2
Physical Form Neat
Linear Formula C22H18O10
MDL Number MFCD22586455
Synonym 3,4,5-Trihydroxybenzoic acid (2 R,3 S)-2-(3,4-dihydroxyphenyl)-3,4-dihydro-3,7-dihydroxy-2 H-1-benzopyran-5-yl ester
Type Analytical Standard
Quantity 10 mg
Formula Weight 442.37
Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.