Learn More
Cayman Chemical (+)-Catechin (hydrate), 225937-10-0, 5 g
SDP
Supplier:  Cayman Chemical 709405
Encompass_Preferred
Catechin is a polyphenolic flavonoid which has been isolated from a variety of natural sources including tea leaves, grape seeds, and the wood and bark of trees such as acacia and mahogany. Catechin is a more potent antioxidant than ascorbate or α-tocopherol in certain in vitro lipid peroxidation assays.{7582} Catechin inhibits the free radical-induced oxidation of isolated LDL by AAPH.{7442} It has also been reported to act as an inhibitor of COX-1 with an IC50 value of about 80 µM.{8140} Catechin and other related procyanidin compounds have antitumor activity when tested in a two-stage mouse epidermal carcinoma model employing topical application.{7582}
Catalog No. NC9854212
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.