Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Catechol O-methyltransferase/COMT, Rabbit, Unlabeled, Polyclonal,

Catalog No. AB5873IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
AB5873IMI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5873IMI Supplier MilliporeSigma™ Supplier No. AB5873I
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Catechol O-methyltransferase (EC 2.1.1.6; UniProt P21964) is encoded by the COMT gene (Gene ID 1312) in human. Catechol-O-methyltransferase initiates the metabolic breakdown of catecholamines (including the neurotransmitters dopamine, epinephrine, and norepinephrine) by catalyzing the transfer of a methyl group from S-adenosylmethionine (SAM) to catecholamines in the presence Mg++. This O-methylation results in one of the major degradative pathways of the catecholamine transmitters. In addition to endogenous substances, COMT is also important in the metabolism of catechol drugs. Two isoforms exist as a result of alternative initiation codon usage. The membrane-bound isoform or MB-COMT contains a short N-terminal cytoplasmic tail (a.a. 1-6), a transmembrane segment (a.a. 7-26), and a large C-terminal extracellular region (a.a. 27-271), while the soluble or S-COMT isoform lacks the N-terminal 50-amino acid sequence of MB-COMT. MB-COMT and S-COMT differ in their substrate affinity and capacities. Despite having a lower capacity than S-COMT, MB-COMT has around a 10-fold greater affinity for dopamine and noradrenaline, and therefore is likely to be better suited for metabolizing catecholamines at the concentrations found in brain. The influence of the Val(158)Met polymorphic form of catechol-o-methyltransferase (COMT) on brain activation and functional connectivity is the subject of numerous studies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Catechol O-methyltransferase/COMT
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene COMT
Gene Accession No. P21964
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to an extracellular domain sequence of human catechol O-methyltransferase.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_000745
Test Specificity This polyclonal antibody was raised against a sequence present in both the membrane-bound (MB-COMT) and the soluble (S-COMT) spliced isoforms of human catechol O-methyltransferase reported by UniProt (P21964).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.