Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Catenin delta-2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144657
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144657 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144657 Supplier Invitrogen™ Supplier No. PA5144657
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Human brain tissue lysate, mouse brain tissue lysate, rat brain tissue lysate. Predicted band size: 133 kDa Subcellular Location: Nucleus, Cell projection, dendrite, Perikaryon.

Armadillo-like proteins are characterized by a series of armadillo repeats, first defined in the Drosophila 'armadillo' gene product, that are typically 42 to 45 amino acids long. These proteins can be divided into subfamilies based on the number of repeats, their overall sequence similarity, and the dispersion of the repeats throughout their sequences. Members of the p120(ctn)/plakophilin subfamily of Armadillo-like proteins, including CTNND1, Catenin delta-2, PKP1, PKP2, PKP4, and ARVCF, have 10 armadillo repeats each (Bonne et al., 1998 [PubMed 9721216]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Catenin delta-2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CTNND2
Gene Accession No. O35116, O35927, Q9UQB3
Gene Alias catenin (cadherin associated protein), delta 2; catenin (cadherin-associated protein), delta 2; catenin (cadherin-associated protein), delta 2 (neural plakophilin-related arm-repeat protein); catenin delta 2; catenin delta-2; catenin delta-2; LOW QUALITY PROTEIN: catenin delta-2; Catnd2; Ctnnd2; Delta-catenin; GT24; Neural plakophilin-related ARM-repeat protein; neurojugin; Neurojungin; Nprap; T-cell delta-catenin
Gene Symbols CTNND2
Host Species Rabbit
Immunogen Recombinant protein within Human Catenin delta-2 1-200 aa.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114028, 1501, 18163
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.