Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Catenin Mouse, Unlabeled, Clone: 14, BD

Catalog No. p-3855693 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610153 50 μg
BDB610154 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610153 Supplier BD Biosciences Supplier No. 610153
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

β-Catenin is a 92kDa protein that binds to the cytoplasmic tail of E-Cadherin. The cadherins, transmembrane adhesion molecules, are found with catenins at adherens junctions (zonula adherens). Deletions in the cytoplasmic domain of E-Cadherin which eliminate catenin binding also result in a loss of cell adhesion, indicating that this binding is essential for E-Cadherin function. Although the α- and β-Catenins have been cloned, very little is known about their biochemical roles. However a link between β-Catenin and colon cancer has been described. β-Catenin was found to co-immunoprecipitate with the APC tumor suppressor protein in human colorectal tumor cell lines, as well as in human kidney 293 cells. E-Cadherin, however, was not detectable in these complexes. Thus the APC-Catenin complex may be affecting the transmission of contact inhibition signals and/or the regulation of cell adhesion.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen β-Catenin
Applications Western Blot
Classification Monoclonal
Clone 14
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse β-Catenin aa. 571-781
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.