Learn More
CPC Scientific H-Ser-Ser-Met-Lys-Leu-Ser-Phe-Arg-Ala-Arg-Ala-Tyr-Gly-Phe-Arg-Gly-Pro-Gly-Pro-Gln-Leu-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific CATS001B
Encompass
Catestatin is a fragment of chromogranin A (parathyroid secretory protein 1), neuroendocrine secretory protein. Catestatin is an inhibitor of catecholamine release. Diminished expression of catestatin might increase the risk for hypertension.ONE-LETTER SEQUENCE: SSMKLSFRARAYGFRGPGPQLMOLECULAR FORMULA: C104H164N32O27S1MOLECULAR WEIGHT:2326.71STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [197151-46-5], RESEARCH AREA: CardiovascularREFERENCES: C.V.Taylor et al., J. Biol. Chem., 275, 22905 (2000); N.R.Mahapatra et al., J. Clin. Invest., 115, 1942 (2005); N.R.Mahapatra, Cardiovasc. Res., 80, 330 (2008)
Catalog No. 50-210-9105
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.