Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cathepsin A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102793
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102793 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102793 Supplier Invitrogen™ Supplier No. PA5102793
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CTSA.

The cathepsin family of proteolytic enzymes include several diverse classes of proteases. Cathepsins B, L, H, K, S and O comprise the cysteine protease class. Cathepsins D and E comprise the aspartyle protease class. The serine protease class includes cathepsin G. Cathepsins function in cellular metabolism and participate in peptide biosynthesis and protein degradation. Cathepsin A, a serine carboxypeptidase, exists in a high molecular weight lysosomal complex with b-galactosidase (b-gal) and a-neuraminidase (Neu1). Cathepsin A functions to protect b-gal and Neu1 from intralysosomal proteolysis. Deficiencies in cathepsin A lead to deficiencies in b-gal and Neu1. The gene encoding human cathepsin A maps to chromosome 20q13.12. Mutations in this gene cause glactosialidosis, a lysosomal storage disorder resulting from the b-gal and Neu1 deficiencies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cathepsin A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CTSA
Gene Accession No. P10619, P16675
Gene Alias AU019505; beta-galactosidase 2; beta-galactosidase protective protein; carboxypeptidase C; Carboxypeptidase L; carboxypeptidase Y-like kininase; carboxypeptidase-L; cathepsin A; CTSA; deamidase; GLB2; GSL; lysosomal carboxypeptidase A; lysosomal protective protein; Lysosomal protective protein 20 kDa chain; Lysosomal protective protein 32 kDa chain; NGBE; PPCA; PPGB; protective protein cathepsin A; Protective protein for beta-galactosidase; urinary kininase
Gene Symbols CTSA
Host Species Rabbit
Immunogen A synthesized peptide derived from human CTSA(Accession P10619), corresponding to amino acid residues L394-F444.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19025, 296370, 5476
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.