Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cathepsin F Polyclonal Antibody, Invitrogen™

Catalog No. PIPA548002
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA548002 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA548002 Supplier Thermo Scientific Supplier No. PA548002

Goat Polyclonal Antibody

In Western blots, approximately 50% cross-reactivity with recombinant mouse Cathepsin F is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Cathepsin F Polyclonal antibody specifically detects Cathepsin F in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cathepsin F
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Ctsf
Gene Accession No. Q9UBX1
Gene Alias AI481912; Cathepsin F; CATSF; CLN13; CTSF; HGNC:2531
Gene Symbols Ctsf
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Cathepsin F Ala20-Asp484
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8722
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.