Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Cathepsin G

Catalog No. ICN19134490 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19134490 500 μg
ICN19134480 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19134490 Supplier MP Biomedicals™ Supplier No. 0219134490

missing translation for 'validMainframeMsg'

From human neutrophils

E.C.3.4.21.20. Enzyme degrades collagen and proteoglycans, and also exhibits releasing activity of angiotensin II from angiotensin I and angiotensinogen. Purity: ≥95% by SDS-PAGE.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2 to 4 μ/mg
Color White
Purity >95% by SDS-Page
For Use With (Application) Act as a bacteriocidal reagent
Content And Storage -70°C
Source Human neutrophils
Quantity 500 μg
Product Type Cathepsin G
Molecular Weight (g/mol) 23500
Inhibitors Chymostatin, DFP
Form Lyophilized Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.