Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Cathepsin G Inhibitor I

Catalog No. AAJ65957MCR
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65957MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ65957MCR Supplier Thermo Scientific Chemicals Supplier No. J65957MCR
Add to Cart

missing translation for 'validMainframeMsg'

 

A potent, selective, reversible, competitive, non-peptide inhibitor of cathepsin G

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 429676-93-7
Molecular Formula C36H33N2O6P

Specifications

Quantity 1 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) A potent, selective, reversible, competitive, non-peptide inhibitor of cathepsin G
Formula Weight 620.60
Sensitivity Light Sensitive
Solubility Soluble in DMSO
Form Solid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.