Learn More
Thermo Scientific Chemicals Cathepsin K Substrate (fluorogenic)
For cellular and molecular biology applications | 578.65 g/mol
$347.50
Specifications
Color | White |
---|---|
Form | Solid |
For Use With (Application) | A sensitive fluorogenic substrate for cathepsin K |
Substrate Type | Cathepsin Substrate |
Molecular Weight (g/mol) | 578.65 |
Catalog Number | Mfr. No. | Substrate Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Substrate Quantity | Price | Quantity & Availability | |||||
AAJ64082MB
|
Thermo Scientific Chemicals
J64082MB |
25mg |
Each of 1 for $347.50
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
A sensitive fluorogenic substrate for cathepsin K
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
White | |
A sensitive fluorogenic substrate for cathepsin K | |
578.65 |
Solid | |
Cathepsin Substrate | |
-30° to -10°C |
Safety and Handling
Recommended Storage : Store at -20°C
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.