Learn More
Montana Molecular Cathepsin-L BacMam - 30mL
SDP
Supplier:  Montana Molecular C1150N030
Encompass_Preferred
Unlabeled Cathepsin-L packaged in BacMam for expression in mammalian cells. BacMam provides even, near 100% transduction efficiency in HEK293 cells as well as expression in many other cell lines, iPSCs, and primary cultures including A549s and HUVECs. Expression is easily titrateable to your experimental needs or to adjust assay sensitivity. This kit includes the Cathepsin-L BacMam and 500mM sodium butyrate in H2O. Use with other Montana Molecular tools including Pseudo-SARS-CoV-2 and BacMam-packaged ACE2, TMPRSS2, and Neuropilin-1.
Catalog No. NC2600311
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.