Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ Cathode Buffer Container Dx

Catalog No. 4408258
Encompass_Preferred
Change view
Click to view available options
Quantity:
4 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-082-58 4 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 44-082-58 Supplier Applied Biosystems™ Supplier No. 4408258
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Cathode Buffer Container Dx contains 1X running buffer to support electrophoresis applications on Applied Biosystems Dx genetic analyzers.

The Cathode Buffer Container Dx is for use with Applied Biosystems Dx genetic analyzers.

The Cathode Buffer Container Dx contains 1X running buffer to support electrophoresis applications on Applied Biosystems Dx genetic analyzers. The Cathode Buffer Container Dx has two separate compartments: the left side provides the cathode buffer for electrophoresis and the right side provides for a capillary wash and spent polymer waste ejection functionality between injections. The Cathode Buffer Container Dx is a ready-to-use, disposable container with a radio frequency identification (RFID) tag incorporated into the label. The top of the Cathode Buffer Container Dx is heat sealed with a plastic film, which should be removed prior to direct installation on to the instrument. Each package includes four individual containers. The Septa Cathode Buffer Container Dx (sold separately) is used for sealing the Cathode Buffer Container Dx container.

This product is intended for in vitro diagnostic use on Applied Biosystems Dx genetic analyzers and is not validated for use with other systems or applications. Applied Biosystems Dx genetic analyzers are to be used by trained operators in the analysis of human DNA for the detection of fluorescently labeled DNA nucleotides by capillary electrophoresis. Refer to the appropriate Applied Biosystems Dx genetic analyzer user guide for further details.

The cathode buffer container (CBC) contains 1x running buffer to support all electrophoresis applications on the 3500 Dx series Genetic Analyzers.

Order Info

4 Pack

TRUSTED_SUSTAINABILITY

Specifications

Starting Material DNA, RNA
Quantity 4 Ea.
For Use With (Equipment) 3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer
Product Type Cathode Buffer Container
Shipping Condition Wet Ice
Content And Storage Each package contains 4 buffer containers.
Store at 2 - 8°C.

For In Vitro Diagnostic Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.