Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CATIP Monoclonal Antibody (2C3)
GREENER_CHOICE

Catalog No. PIMA523285
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA523285 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA523285 Supplier Invitrogen™ Supplier No. MA523285
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MSSKVYSTGS RAKDHQPSGP ECLPLPEANA EAIDFLSSLH KEELQMLFFS ETLAMVSDTG EPQGELTIEV QRGKYQEKLG MLTYCLFVHA SSRGFLDKML CGNSLLGYLS EKLELMEQHS QDFIKFLILP MERKMSLLKQ DDQLAVTRSI KEGEEVKTGV TSFPWSSIKG FISEAANLVL LRVMAWRRMV PSNARFLTLD TEGKLCYLTY QNLGFQTIQV DHQQAEVFIV EQTVHAEEGI PMSCQYYLLS DGHLAKRIQV GSPGCCIITK MPILREEDEI EPRPVFEKKP LVWEEDMELY SKFLDRKEEL RLGHASYLRQ HPEAHALISD FLLFLLLRQP EDVVTFAAEF FGPFDPWRPS SPALGSSHRP NPFRSLEPEG DARSGAA.

Plays a role in primary ciliogenesis by modulating actin polymerization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CATIP
Applications ELISA
Classification Monoclonal
Clone 2C3
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene CATIP
Gene Accession No. Q7Z7H3
Gene Alias C15H2orf62; C2orf62; C37H2orf62; C6H2orf62; Catip; ciliogenesis associated TTC17 interacting protein; Ciliogenesis-associated TTC17-interacting protein; Gm216; hypothetical protein LOC690437; LOC690437
Gene Symbols CATIP
Host Species Mouse
Immunogen CATIP (AAH52750, 1 a.a. ~ 387 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 375307
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.