Learn More
Strem, An Ascensus Company CAS 13965-03-2. 25 x 1vial. trans-Dichlorobis(triphenylphosphine)palladium(II)/potassium phosphate admixture [CatKit single-use vials - 6.32 wt% Pd complex]. MFCD18827649
SDP
Supplier:  Strem, An Ascensus Company 46203825VL
Encompass_Preferred
CAS 13965-03-2. 25 x 1vial. trans-Dichlorobis(triphenylphosphine)palladium(II)/potassium phosphate admixture [CatKit single-use vials - 6.32 wt% Pd complex]. MFCD18827649. Molecular Weight 701.89. Molecular Formula PdCl2[P(C6H5)3]2/K3PO4. Color/form off-white pwdr. Strem 46-2038. http//www.strem.com/catalog/v/46-2038/
Catalog No. 50-211-0847
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.