Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CaV2.3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674818
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-818 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-818 Supplier Invitrogen™ Supplier No. PA112664
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CaV2.3 Polyclonal Antibody for Western Blot

The expression of multiple classes of voltage dependent calcium channels (VDCCs) allows neurons to tailor calcium signaling to functionally discrete cellular regions. Although N-Type VDCCs exist before birth which is consistent with a role in migration, most N-Type VDCCs subunit expression is postnatal and is important in the genesis of synaptic transmission in discrete hippocampal fields.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CaV2.3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene Cacna1e
Gene Accession No. Q15878
Gene Alias A430040I15; alpha1E; BII; brain calcium channel II; CACH6; CACHA1E; CACNA1E; Cacnl1a6; calcium channel, L type, alpha-1 polypeptide, isoform 6; calcium channel, R type, alpha-1 polypeptide, isoform 6; calcium channel, voltage-dependent, L type, alpha 1E subunit; calcium channel, voltage-dependent, R type, alpha 1E subunit; calcium voltage-gated channel subunit alpha1 E; Cav2.3; Cchra1; gm139; RBE2; RBE-II; voltage gated calcium channel alpha1E subunit; voltage-dependent calcium channel alpha 1E subunit; voltage-dependent calcium channel alpha1-E subunit; voltage-dependent R-type calcium channel subunit alpha-1E; voltage-gated calcium channel alpha subunit Cav2.3; voltage-gated calcium channel subunit alpha Cav2.3
Gene Symbols Cacna1e
Host Species Rabbit
Immunogen Synthetic peptide, conjugated to KLH, corresponding to the rat alpha1E calcium channel.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 777
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.