Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Caveolin 1 Polyclonal Antibody

Catalog No. 01673183
Change view
Click to view available options
Quantity:
500 μL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-673-183 1 mL
PIPA532297 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-673-183 Supplier Invitrogen™ Supplier No. PA137260
  • $695.00
    $570.30 / Each of 1

    Save $124.70

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is lung carcinoma.

Caveolin-1 or CAV1 is a scaffolding proteins within caveolar membranes. It interacts directly with G-protein alpha subunits and can functionally regulate their activity. Caveolin-1 is also involved in the co-stimulatory signal essential for T-cell receptor (TCR) mediated T-cell activation. It can bind with DPP4 and induces T-cell proliferation and NF-kappa-B activation in a T-cell receptor/CD3-dependent manner. Mutations affecting the CAV1 gene can result in congenital generalized lipodystrophy 3, pulmonary hypertension primary 3, or partial lipodystrophy/congenital cataracts, and neurodegeneration syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caveolin 1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.1% sodium azide; pH 7.6
Gene CAV1
Gene Accession No. Q03135
Gene Alias BSCL3; CAV; CAV1; Cav-1; caveolae; caveolin 1; caveolin 1, caveolae protein; caveolin 1, caveolae protein, 22kDa; caveolin 1a; caveolin 1b; caveolin caveolae protein 22 kDa; caveolin, caveolae protein 1; caveolin, caveolae protein, 22 kDa; caveolin-1; caveolin-1a; caveolin-1b; cell growth-inhibiting protein 32; CGL3; HGNC:1527; LCCNS; MSTP085; OTTHUMP00000196475; PPH3; Unknown (protein for IMAGE:7985573); variant a; variant b; vesicular integral-membrane protein VIP21; VIP21; wu:fc07c04
Gene Symbols CAV1
Host Species Rabbit
Immunogen Synthetic peptide from N-terminus of human caveolin-1.
Purification Method Antigen affinity chromatography
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 857
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.