Learn More
Cayman Chemical CAY10499, 359714-55-9, 1 mg
SDP
Supplier:  Cayman Chemical 100078751
Encompass_Preferred
A non-selective lipase inhibitor (IC50s = 144, 90, and 14 nM for human recombinant MAGL, HSL, and FAAH, respectively); inhibits ATGL, DAGLa, ABHD6, and CES1 at 5 mM; inhibits the growth of MCF-7, MDA-MB-231, COV318, and OVCAR-3 cancer cells (IC50s = 4.2, 46, 106.7, and 79.8 mM, respectively); reduces FRUC diet- and AIN-93M diet-induced increases in cytosolic lipase activity in rats
Catalog No. NC1176233
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.