Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam CAY10585, HIF-1alpha Inhibitor, Solid, 5 mg

Catalog No. 502705759
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5759 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5759 Supplier Abcam Supplier No. AB1444225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent HIF-1α inhibitor with >97% purity that suppresses transcription of target genes VEGF and erythropoietin.

  • Molecular Weight: 435.5 Da
  • Purity: >97%
  • Mechanism: HIF-1α inhibitor (IC50 values are 2.6 μM for Hep3B and 0.7 μM for AGS cell lines)
  • Biological activity: Suppresses transcription of HIF-1 target genes VEGF and erythropoietin at 10 μM; attenuates hypoxia-induced ANP secretion; blunts BMP9-induced osteogenic signaling in MSCs
  • Form: Solid

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Target Molecule HIF-1α
Substrate Properties Chemical Substrate
Alternate Part Number CAY10585
Content And Storage Store at -20°C.
Form Solid
Quantity 5 mg
For Use With (Application) HIF-1α inhibition, transcription suppression, osteogenic signaling inhibition
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.