Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam CAY10621 (SKI 5C), SPHK 1 inhibitor, 5 mg

Catalog No. 502705781
Encompass_Preferred
Catalog No. 50-270-5781 Supplier Abcam Supplier No. AB1444105MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAY10621 (SKI 5C), inhibitor.

  • Class: Inhibitor
  • Form: Liquid
  • Purity: >98%
  • Molecular Weight: 435.6 g/mol

Specifications

Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.