Learn More
Apexbio Technology LLC CB-839 10mg. 685898-44-6. MFCD11840749
SDP
Supplier:  Apexbio Technology LLC B479910
Encompass_Preferred
CB-839(Telaglenastat) is a potent, selective, and orally bioavailable glutaminase inhibitor with IC50 of 24 nM for recombinant human GAC(glutaminase C). CB-839 has antiproliferative activity in a triple-negative breast cancer (TNBC) cell line, HCC-1806, while no antiproliferative activity is observed in an estrogen receptor–positive cell line, T47D. In the mouse JIMT-1 xenograft model, CB-839 alone (200 mg/kg, p.o.) results in 54% tumor growth inhibition (TGI) relative to vehicle control, combination of CB-839 (200 mg/kg, p.o.) with paclitaxel (10 mg/kg, p.o.) largely suppresses the regrowth of the tumors resulting in a TGI relative to vehicle control of 100%. Other sizes are available. Please inqury us for quote.
Catalog No. 50-115-1911
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.