Learn More
Enzo Life Sciences Cannabinoid receptor CB1 polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905708100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the N-terminus of mouse cannabinoid receptor CB1. Species reactivity: Human, Mouse, Rat Chicken. Applications: ELISA, IHC, WB. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.01% sodium azide.
Catalog No. 50-200-7001
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.