Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ CBL 2
SDPSCI_ED

Catalog No. S15991ND Shop All Vernier Software Products
Change view
Click to view available options
Sensor Types:
3 analog and 1 digital
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sensor Types
S15991ND 3 analog and 1 digital
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15991ND Supplier Vernier™ Supplier No. CBL2

missing translation for 'validMainframeMsg'

Includes

CBL 2 interface, Stainless steel temperature probe, Light probe, Voltage probe, Calculator cradle and link cable, User's guide, Ordering Information, This product is discontinued and no longer available.

The CBL 2 is a low-cost interface for Texas Instruments graphing calculators such as the TI-84 Plus.

  • For classrooms that with wide availability of compatible TI graphing calculators, the CBL 2 is an inexpensive way to collect sensor data.
  • The CBL 2 is portable and battery powered, you can take it out of the classroom for experiments such as measuring accelerations on amusement rides or monitoring the temperature, dissolved oxygen, and pH of a lake or stream.
  • CBL 2 is not compatible with TI-84 Plus CE or TI-Nspire Handhelds.
  • Includes:
    • CBL 2 interface
    • Stainless steel temperature probe
    • Light probe
    • Voltage probe
    • Calculator cradle and link cable
    • User’s guide

Warranty and Services

1 year warranty from Texas Instruments

Specifications

Type Interface
Description Stores up to 12,000 data points
Includes CBL 2 interface, stainless steel temperature, light probe, voltage probe, calculator cradle and link cable, users quide
Grade Middle School and High School
Interface CBL 2 interface
Sensor Types 3 analog and 1 digital
Warranty 1-year limited warranty from Texas Instruments
For Use With (Application) Educational use only
Rate 50,000 readings per second
Resolution 10-bit A/D convrsion
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.