Learn More
Cable Ties And More CBL TIES 7.4INCHES 50LBS 100PC
SDP
Supplier:  Cable Ties And More TF07
Encompass
Panduit Tefzel Zip Ties FeaturesMaterial - Ethylene-tetrafluoroethylene Highly Resistant to Concentrated AcidsGood Radiation Resistance up to 100 MegaradsRoHS CompliantWeather ResistantUV Resistant UV Resistant- For indoor or out door useMax Operating Temp 302 degrees F / 150 degrees CMeets the requirements of IEEE 383
Catalog No. NC2752723
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.