Learn More
Cayman Chemical CBL0137, 1197996-80-7, 1 mg
Supplier: Cayman Chemical 191101
Molecular Formula C21H24N2O2, Purity ≥95%, Formula Weight 336.4, Synonyms: 1,1'-[9-[2-[(1-methylethyl)amino]ethyl]-9H-carbazole-3,6-diyl]bis-ethanone, CBLC137, Curaxin 137; A metabolically stable curaxin that activates p53 with an EC50 value of 0.37 M and inhibits NF-B with an EC50 of 0.47 M; has broad anticancer activity in mice when administered orally.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.