Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CBLB (Human) Recombinant Protein

Catalog No. 89998072 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-072 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-072 Supplier Abnova™ Supplier No. H00000868P01
Add to Cart
Add to Cart

Human CBLB full-length ORF ( AAH32851.1, 1 a.a. - 982 a.a.) recombinant protein with GST-tag at N-terminal.

  • Sequence: MANSMNGRNPGGRGGNPRKGRILGIIDAIQDAVGPPKQAAADRRTVEKTWKLMDKVVRLCQNPKLQLKNSPPYILDILPDTYQHLRLILSKYDDNQKLAQLSENEYFKIYIDSLMKKSKRAIRLFKEGKERMYEEQSQDRRNLTKLSLIFSHMLAEIKAIFPNGQFQGDNFRITKADAAEFWRKFFGDKTIVPWKVFRQCLHEVHQISSGLEAMALKSTIDLTCNDYISVFEFDIFTRLFQPWGSILRNWNFLAVTHPGYMAFLTYDEVKARLQKYSTKPGSYIFRLSCTRLGQWAIGYVTGDGNILQTIPHNKPLFQALIDGSREGFYLYPDGRSYNPDLTGLCEPTPHDHIKVTQEQYELYCEMGSTFQLCKICAENDKDVKIEPCGHLMCTSCLTAWQESDGQGCPFCRCEIKGTEPIIVDPFDPRDEGSRCCSIIDPFGMPMLDLDDDDDREESLMMNRLANVRKCTDRQNSPVTSPGSSPLAQRRKPQPDPLQIPHLSLPPVPPRLDLIQKGIVRSPCGSPTGSPKSSPCMVRKQDKPLPAPPPPLRDPPPPPPERPPPIPPDNRLSRHIHHVESVPSKDPPMPLEAWCPRDVFGTNQLVGCRLLGEGSPKPGITASSNVNGRHSRVGSDPVLMRKHRRHDLPLEGAKVFSNGHLGSEEYDVPPRLSPPPPVTTLLPSIKCTGPLANSLSEKTRDPVEEDDDEYKIPSSHPVSLNSQPSHCHNVKPPVRSCDNGHCMLNGTHGPSSEKKSNIPDLSIYLKGDVFDSASDPVPLPPARPPTRDNPKHGSSLNRTPSDYDLLIPPLGEDAFDALPPSLPPPPPPARHSLIEHSKPPGSSSRPSSGQDLFLLPSDPFVDLASGQVPLPPARRLPGENVKTNRTSQDYDQLPSCSDGSQAPARPPKPRPRRTAPEIHHRKPHGPEAALENVDAKIAKLMGEGYAFEEVKRALEIAQNNVEVARSILREFAFPPPVSPRLNL

Specifications

Accession Number AAH32851.1
Gene ID (Entrez) 868
Name Cas-Br-M (murine) ecotropic retroviral transforming sequence b
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
Quantity 2 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias DKFZp686J10223, DKFZp779A0729, DKFZp779F1443, FLJ36865, FLJ41152, Nbla00127, RNF56
Gene Symbol CBLB
Species Wheat Germ (in vitro)
Protein Tag GST
Buffer 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.