Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBLB Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2D3A2, Proteintech
SDP

Catalog No. p-7213319 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-114-2721 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-114-2721 Supplier Proteintech Group Inc Supplier No. 663531IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene encodes a protein of unknown function. The protein contains a bromodomain, a sequence motif often found in transcriptional coactivators, and localizes to the nucleus in testis and several other cell types.

Specifications

Antigen CBLB
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2D3A2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CBLB
Gene Accession No. Q13191, Q3TTA7, Q8K4S7
Gene Alias CBLB, Nbla00127, RING finger protein 56, RNF56, SH3 binding protein CBL B
Gene Symbols Cblb
Host Species Mouse
Immunogen CBLB Fusion Protein Ag18757
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100621690, 171136, 208650, 868
Target Species Human, Mouse, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.